missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody (6D4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26582-100ul
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| FGFR3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 6D4 | |
| Western Blot 1:500-1:5000, ELISA | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| A synthesized peptide derived from Human FGFR3 [UniProt P22607] | |
| 100 μL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction