missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (FB-8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1128-0.025ml
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Bovine samples. It is validated for Western Blot, ELISA, Radioimmunoassay, Block/Neutralize.
Specifications
| FGF basic/FGF2 | |
| Monoclonal | |
| 2.4 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Blocking/Neutralizing 5 μg/mL | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Does not cross-react with human fibroblast growth factor - acidic (aFGF). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| FB-8 | |
| Unconjugated | |
| P09038 | |
| FGF2 | |
| Recombinant full length protein (Human). | |
| 0.025 mL | |
| Angiogenesis, Cancer | |
| 2247 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction