missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (FB-8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
191.00€
Specifications
| Antigen | FGF basic/FGF2 |
|---|---|
| Clone | FB-8 |
| Concentration | 2.4 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Blocking/Neutralizing 5 μg/mL |
| Applications | Western Blot |
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Bovine samples. It is validated for Western Blot, ELISA, Radioimmunoassay, Block/Neutralize.Specifications
| FGF basic/FGF2 | |
| 2.4 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| P09038 | |
| 2247 | |
| Recombinant full length protein (Human). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| FB-8 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Blocking/Neutralizing 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| FGF2 | |
| IgG1 | |
| Protein A purified | |
| Does not cross-react with human fibroblast growth factor - acidic (aFGF). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title