missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Antibody (12H1), Novus Biologicals™
Mouse Monoclonal Antibody
355.00€ - 568.00€
Specifications
| Antigen | Ferritin |
|---|---|
| Clone | 12H1 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043945
|
Novus Biologicals
NBP3-26568-50ul |
50 μL |
355.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043138
|
Novus Biologicals
NBP3-26568-100ul |
100 μL |
568.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| Ferritin | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant Human Ferritin protein [UniProt P02794] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 12H1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 2495 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title