missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Antibody (12H1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26568-100ul
This item is not returnable.
View return policy
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Ferritin | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 12H1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant Human Ferritin protein [UniProt P02794] | |
| 100 μL | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 2495 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction