missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty Acid Synthase/FASN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
237.00€ - 531.00€
Specifications
| Antigen | Fatty Acid Synthase/FASN |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500immunoprecipitation 1:100, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18738023
|
Novus Biologicals
NB400-114SS |
0.025 mL |
237.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18715273
|
Novus Biologicals
NB400-114 |
531.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Fatty Acid Synthase/FASN Polyclonal specifically detects Fatty Acid Synthase/FASN in Human, Mouse, Rat, Porcine, Chicken, Feline, Hamster, Plant, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Fatty Acid Synthase/FASN | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology | |
| Whole antisera with 0.05% Sodium Azide | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| FASN | |
| IgG | |
| Unpurified | |
| 272 kDa |
| Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500immunoprecipitation 1:100, Knockdown Validated | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Pig, Chicken, Feline, Hamster, Plant, Primate | |
| P19096 | |
| 2194 | |
| A synthetic peptide, conjugated to KLH, made near the N-terminus of mouse FAS. [Swiss-Prot# P19096] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title