missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty Acid Synthase/FASN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
Brand: Novus Biologicals NB400-114SS
This item is not returnable.
View return policy
Description
Fatty Acid Synthase/FASN Polyclonal specifically detects Fatty Acid Synthase/FASN in Human, Mouse, Rat, Porcine, Chicken, Feline, Hamster, Plant, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Fatty Acid Synthase/FASN | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500immunoprecipitation 1:100, Knockdown Validated | |
| P19096 | |
| FASN | |
| A synthetic peptide, conjugated to KLH, made near the N-terminus of mouse FAS. [Swiss-Prot# P19096] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Chicken, Feline, Hamster, Plant, Primate | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| Rabbit | |
| 272 kDa | |
| 0.025 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology | |
| 2194 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction