missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM212A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
220.00€ - 326.00€
Specifications
| Antigen | FAM212A |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654905
|
Novus Biologicals
NBP3-05938-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652456
|
Novus Biologicals
NBP3-05938-50ul |
50 μg |
220.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM212A Polyclonal antibody specifically detects FAM212A in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| FAM212A | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH7.0 | |
| 389119 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C3orf54, chromosome 3 open reading frame 54, family with sequence similarity 212, member A, INKA1, INKA1 protein, MGC20416 | |
| Produced in rabbits immunized with E. coli-derived Human FAM212A fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title