missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP9/T-FABP Antibody (13F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31200-0.1ML
This item is not returnable.
View return policy
Description
FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.
Specifications
| FABP9/T-FABP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:1000 | |
| NP_001073995 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 646480 | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1b κ |
| Western Blot, ELISA | |
| 13F9 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP | |
| Recombinant human FABP9 (1-132aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human FABP9 mAb, clone 13F9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP9 protein. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction