missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP9/T-FABP Antibody (13F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
353.00€ - 468.00€
Specifications
| Antigen | FABP9/T-FABP |
|---|---|
| Clone | 13F9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:1000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18182974
|
Novus Biologicals
NBP2-31200-0.05ML |
0.05 mL |
353.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18145094
|
Novus Biologicals
NBP2-31200-0.1ML |
0.1 mL |
468.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.Specifications
| FABP9/T-FABP | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001073995 | |
| 646480 | |
| IgG1b κ | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 13F9 | |
| Western Blot 1:1000, ELISA 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP | |
| Recombinant human FABP9 (1-132aa) purified from E. coli | |
| Primary | |
| Protein A purified | |
| Anti-human FABP9 mAb, clone 13F9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP9 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title