missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (3C12), Novus Biologicals™
Mouse Monoclonal Antibody
231.00€ - 792.00€
Specifications
| Antigen | Ezrin |
|---|---|
| Clone | 3C12 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203784
|
Novus Biologicals
NB600-1310-0.1ML |
0.1 mL |
231.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18283792
|
Novus Biologicals
NB600-1310 |
0.5 mL |
792.00€
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ezrin Monoclonal specifically detects Ezrin in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Ezrin | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| EZR | |
| IgG1 | |
| Ascites | |
| This antibody reacts with a protein of 81 kDa. Ezrin serves as a tyrosine kinase substrate and is phosphorylated in EGF-stimulated cells. Ezrin is a cytoplasmic protein, enriched in microvilli and other cell surfaces. Ezrin has an actin-binding capacity. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. |
| 3C12 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine, Mouse, Human, Primate, Rat | |
| 7430 | |
| BALB/C mice were immunized with a recombinant fragment of human ezrin corresponding to amino acid 362-585. | |
| Primary | |
| Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title