missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (3C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1310
This item is not returnable.
View return policy
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| Ezrin | |
| Monoclonal | |
| Unconjugated | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| This antibody reacts with a protein of 81 kDa. Ezrin serves as a tyrosine kinase substrate and is phosphorylated in EGF-stimulated cells. Ezrin is a cytoplasmic protein, enriched in microvilli and other cell surfaces. Ezrin has an actin-binding capacity. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3C12 | |
| Western Blot 1:100-1:750, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:100 | |
| EZR | |
| BALB/C mice were immunized with a recombinant fragment of human ezrin corresponding to amino acid 362-585. | |
| 0.5 mL | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| 7430 | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction