Learn More
Ethylene Glycol in DSMO, SPEX CertiPrep™
Product Code. 16349757 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 16349757
Brand: SPEX Certiprep USPS1952DMSO
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Ethylene glycol107-21-13100μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL Amber Prescored Glass Ampule
CAS 107-21-1
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Explosion Limit 1.8% (volume); 63% (volume)
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names Ethylene Glycol
Density 1.1g/cm3
Physical Form Fluid
Molecular Formula C2H6O2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES C(CO)O
Element Ethylene Glycol
IUPAC Name ethane-1,2-diol
Solvent or Matrix Dimethyl Sulfoxide
Molecular Weight (g/mol) 62.068
PubChem CID 174
Quantity 1 mL
ChEBI CHEBI:30742
Grade USP
Concentration or Composition (by Analyte or Components) 3100μg/mL
Chemical Name or Material Ethylene Glycol in Dimethyl Sulfoxide, USP 467 Standard
Show More Show Less
compliance-icons
Product Identifier
  • Stock VOA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : dimethyl sulfoxide
Product intended for laboratory use only