missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethyl Red

Product Code. 11433937
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433937 10 g 10g
11443937 50 g 50g
11458287 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433937 Supplier Thermo Scientific Chemicals Supplier No. A18367.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76058-33-8 | C17H18N3O2 | 296.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76058-33-8
Molecular Formula C17H18N3O2
Molecular Weight (g/mol) 296.35
MDL Number MFCD00002427
InChI Key HBRCDTRQDHMTDA-UHFFFAOYSA-M
Synonym 2-(4-Diethylaminophenylazo)benzoic acid
PubChem CID 54832
SMILES CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O

Specifications

Melting Point ∼135°C (decomposition)
Beilstein 3366056
IUPAC Name 2-[[4-(diethylamino)phenyl]diazenyl]benzoic acid
Formula Weight 297.36
Quantity 10 g
Chemical Name or Material Ethyl Red

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.