CAS RN 76058-33-8
CAS RN 76058-33-8
IUPAC Name:
2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Synonyms:
2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol):
296.35
Molecular Formula:
C17H18N3O2
InChi Key:
HBRCDTRQDHMTDA-UHFFFAOYSA-M
SMILES:
CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ethyl Red
CAS: 76058-33-8 Molecular Formula: C17H18N3O2 Molecular Weight (g/mol): 296.35 MDL Number: MFCD00002427 InChI Key: HBRCDTRQDHMTDA-UHFFFAOYSA-M Synonym: 2-(4-Diethylaminophenylazo)benzoic acid PubChem CID: 54832 SMILES: CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O