Learn More
Ethyl 4-cyano-1H-pyrazole-5-carboxylate, 95%, Thermo Scientific Chemicals
Product Code. 15407417
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15407417
Brand: Thermo Scientific Alfa Aesar H34460.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 318497-88-0
Molecular Formula C7H7N3O2
Molecular Weight (g/mol) 165.152
MDL Number MFCD00664125
InChI Key COJOZBXPRZQJLK-UHFFFAOYSA-N
Synonym ethyl 4-cyano-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 4-cyano-, ethyl ester, ethylcyanopyrazolecarboxylate, 4-cyano-2h-pyrazole-3-carboxylic acid ethyl ester
PubChem CID 1479416
IUPAC Name ethyl 4-cyano-1H-pyrazole-5-carboxylate
SMILES CCOC(=O)C1=C(C=NN1)C#N
Specifications
Melting Point 226°C to 228°C
Color Orange to White
Quantity 250 mg
Formula Weight 165.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ethyl 4-cyano-1H-pyrazole-5-carboxylate