Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Product Code. 15148354 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
12 x 5 mL Eppendorf Tubes
24 Spin Columns
24 x 1.5 to 2.0 mL Tubes
24 x 1.5/2 mL
30 x 1.5 to 2.0 mL Tubes
48 x 0.2 mL PCR Tubes
48 x 1.5 to 2.0 mL Tubes
Includes:
Aerosol-tight with Rotor Lid
Plastic Rotor Lid
Rotor Lid
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Includes Unit Size
15148354 12 x 5 mL Eppendorf Tubes Aerosol-tight with Rotor Lid Each
13443396 24 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
13473396 30 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
13453396 48 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
13483396 24 Spin Columns Aerosol-tight with Rotor Lid Each
13463396 30 x 1.5 to 2.0 mL Tubes Rotor Lid Each
13493396 48 x 1.5 to 2.0 mL Tubes Rotor Lid Each
13403406 48 x 0.2 mL PCR Tubes Plastic Rotor Lid Each
13413406 24 x 1.5/2 mL Aerosol-tight with Rotor Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15148354 Supplier Eppendorf™ Supplier No. 5409700006
 Restricted Item
Estimated Shipment: 16-12-2026
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Type Aerosol-tight Rotor
Capacity 12 x 5 mL Eppendorf Tubes
Max. RCF 20,598 x g
Max. Speed 14,000 rpm
For Use With (Equipment) Eppendorf QuickLock™ 5427 R
Includes Aerosol-tight with Rotor Lid
No. of Places 12
Model FA-45-12-17