missing translation for 'onlineSavingsMsg'
Learn More

EME1 Mouse anti-Human, Clone: MTA31 7H2/1, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen EME1
Clone MTA31 7H2/1
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1-2 ug per 100-500 μg of protein 1ml of lysate, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18625566
View Documents
Novus Biologicals
NBP3-07779-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EME1 Monoclonal specifically detects EME1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

EME1
0.2 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
DNA Repair, Homologous Recombination
10 mM PBS with 0.05% BSA
146956
IgG1 κ
Protein A or G purified
MTA31 7H2/1
Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1-2 ug per 100-500 μg of protein 1ml of lysate, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A
His-tagged recombinant EME1 of human origin
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.