missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EME1 Mouse anti-Human, Clone: MTA31 7H2/1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07779-100ug
This item is not returnable.
View return policy
Description
EME1 Monoclonal specifically detects EME1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| EME1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1-2 ug per 100-500 μg of protein 1ml of lysate, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A | |
| His-tagged recombinant EME1 of human origin | |
| 100 μg | |
| DNA Repair, Homologous Recombination | |
| 146956 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| MTA31 7H2/1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction