missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
189.00€
Specifications
| Antigen | Dynamin 2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Dynamin 2 Polyclonal specifically detects Dynamin 2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.Specifications
| Dynamin 2 | |
| Western Blot 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| 1785 | |
| Synthetic Peptide: C S(760) P T P Q R R P V S S I H P P G R P P A(779) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
| DNM2 | |
| IgG | |
| Affinity Purified | |
| Dynamin 2 - Synapse |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title