missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB300-617-0.025ml
This item is not returnable.
View return policy
Description
Dynamin 2 Polyclonal specifically detects Dynamin 2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.
Specifications
| Dynamin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| DNM2 | |
| Synthetic Peptide: C S(760) P T P Q R R P V S S I H P P G R P P A(779) | |
| 0.025 mL | |
| Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience | |
| 1785 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Dynamin 2 - Synapse | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction