Learn More
DWK Life Sciences DURAN™ PURE Bottle, Amber, with DIN Thread
BIOPHARMA
Product Code. p-7189161 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Pack of 12
Pack of 120
Pack of 210
Pack of 280
Pack of 30
Pack of 360
Pack of 48
Pack of 540
Pack of 940
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15565360 25 mL Pack of 940
15310604 25 mL Pack of 10
15575360 100 mL Pack of 540
15320604 100 mL Pack of 10
15585360 250 mL Pack of 280
15330604 250 mL Pack of 10
15336798 500 mL Pack of 360
15340604 500 mL Pack of 10
15350604 1000 mL Pack of 10
15346798 1000 mL Pack of 210
15356798 2000 mL Pack of 120
15360604 2000 mL Pack of 10
15896931 5000 mL Each
15508441 5000 mL Pack of 48
15518441 10,000 mL Pack of 30
15806941 10,000 mL Each
15528441 20,000 mL Pack of 12
15816941 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15565360 Fornitore DWK Life Sciences N. del fornitore 818061406
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
The DURAN™ PURE Bottle, Amber, with DIN Thread is a high-quality Type I borosilicate glass bottle designed for pharmaceutical and biotech applications. Amber colouring protects light-sensitive compounds, and a protective cover prevents contamination during storage and transport.
  • Type I borosilicate glass construction for superior chemical resistance and durability
  • Amber colouring provides reliable protection for light-sensitive compounds
  • Protective cover prevents contamination inside the bottle during storage and transport
  • Available with GL 25 and GL 45 DIN thread closures; sizes from 25 mL to 20 L
  • Certificate of Analysis (CoA) provided with each delivery
  • 100% visual inspection and single batch delivery
  • Hydrolytic resistance tested per EP, USP and JP for each batch
  • 100% traceability of all manufacturing steps
  • Change control management with customer notification in case of changes
  • Drug Master File available; meets requirements of international pharmacopoeias
Specifica
Capacity (Metric) 25 mL
Color Amber
For Use With (Application) Manufacturing of light-sensitive pharmaceutical drugs and biotech products; Storage and transport of light-sensitive APIs, bulk drug intermediates, ingredients and media
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 25
Closure Included No
Graduated Yes
Mouth Narrow
Diameter (Metric) 36 mm
Autoclavable Autoclavable
Capacity (English) 1 oz.
Shape Round
Disposable Reusable
Product Type Screw Neck Bottle
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.