missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dre recombinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
194.00€ - 436.00€
Specifications
| Antigen | Dre recombinase |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:100 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604299
|
Novus Biologicals
NBP3-11868-25ul |
25 μg |
194.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671867
|
Novus Biologicals
NBP3-11868-100ul |
100 μg |
436.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dre recombinase Polyclonal antibody specifically detects Dre recombinase in Nonspecies specific samples. It is validated for Western Blot, ELISASpecifications
| Dre recombinase | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Non-species specific | |
| Polyclonal antibody raised in rabbit against Dre recombinase using a KLH-conjugated synthetic peptide located at the C-terminal part of the protein. | |
| Whole Antiserum |
| Western Blot 1:500, ELISA 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antiserum | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title