missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dre recombinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11868-100ul
This item is not returnable.
View return policy
Description
Dre recombinase Polyclonal antibody specifically detects Dre recombinase in Nonspecies specific samples. It is validated for Western Blot, ELISA
Specifications
| Dre recombinase | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:100 | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Non-species specific | |
| Antisera |
| Western Blot, ELISA | |
| Unconjugated | |
| Whole antiserum | |
| Polyclonal antibody raised in rabbit against Dre recombinase using a KLH-conjugated synthetic peptide located at the C-terminal part of the protein. | |
| 100 μg | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction