missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR3/TNFRSF25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€
Specifications
| Antigen | DR3/TNFRSF25 |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.Specifications
| DR3/TNFRSF25 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| PBS with 0.02% Sodium Azide | |
| Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1 | |
| TNFRSF25 | |
| IgG | |
| Ion exchange chromatography | |
| DR3 has no cross reaction to other death receptors. |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAQ88676 | |
| 8718 | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human DR3. The immunogen is located within the first 50 amino acids of DR3. Amino Acid Squence: QGGTRSPRCDCAAGDFH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 59 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title