missing translation for 'onlineSavingsMsg'
Learn More

DR3/TNFRSF25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76474-0.025mg

 View more versions of this product

Product Code. 18543512

  • 189.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DR3/TNFRSF25
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1
Rabbit
59 kDa
0.025 mg
Apoptosis, Cell Biology
8718
Human
IgG
Western Blot
1 mg/ml
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
AAQ88676
TNFRSF25
Antibody was raised against a 16 amino acid peptide near the amino terminus of human DR3. The immunogen is located within the first 50 amino acids of DR3. Amino Acid Squence: QGGTRSPRCDCAAGDFH
Ion exchange chromatography
RUO
Primary
DR3 has no cross reaction to other death receptors.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only