missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNase II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
171.00€
Specifications
| Antigen | DNase II |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DNase II Polyclonal specifically detects DNase II in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DNase II | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| PBS with 0.02% Sodium Azide | |
| Acid DNase, Deoxyribonuclease II alpha, deoxyribonuclease II, lysosomal, deoxyribonuclease-2-alpha, DNase II alpha, DNase II, lysosomal, DNASE2A, DNL, DNL2, EC 3.1.22.1, Lysosomal DNase II, R31240_2 | |
| DNASE2 | |
| IgG | |
| Affinity Purified | |
| An approximate 40 kDa band can be detected, which represents the pro-enzyme of DNase II. |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| O00115 | |
| 1777 | |
| Antibody was raised against a 14 amino acid peptide near the carboxy terminus of human DNase II. The immunogen is located within the last 50 amino acids of DNase II. Amino Acid Squence: CNGMARKPSRAYKI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| ∼40 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title