missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNase II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76971-0.025mg
This item is not returnable.
View return policy
Description
DNase II Polyclonal specifically detects DNase II in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DNase II | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Acid DNase, Deoxyribonuclease II alpha, deoxyribonuclease II, lysosomal, deoxyribonuclease-2-alpha, DNase II alpha, DNase II, lysosomal, DNASE2A, DNL, DNL2, EC 3.1.22.1, Lysosomal DNase II, R31240_2 | |
| Rabbit | |
| ∼40 kDa | |
| 0.025 mg | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 1777 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 5 μg/mL | |
| O00115 | |
| DNASE2 | |
| Antibody was raised against a 14 amino acid peptide near the carboxy terminus of human DNase II. The immunogen is located within the last 50 amino acids of DNase II. Amino Acid Squence: CNGMARKPSRAYKI | |
| Affinity Purified | |
| RUO | |
| Primary | |
| An approximate 40 kDa band can be detected, which represents the pro-enzyme of DNase II. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction