missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35462-100ul
This item is not returnable.
View return policy
Description
DHX8 Polyclonal antibody specifically detects DHX8 in Human samples. It is validated for ELISA,Western Blot
Specifications
| DHX8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DDX8ATP-dependent RNA helicase DHX8, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase), DEAH (Asp-Glu-Ala-His) box polypeptide 8, DEAH box protein 8, DEAH-box protein 8, EC 3.6.1, EC 3.6.4.13, HRH1RNA helicase HRH1, PRP22, PRPF22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1001-1220 of human DHX8 (NP_004932.1).,, Sequence:, CSEEMLTIVSMLSVQNVFYRPKDKQALADQKKAKFHQTEGDHLTLLAVYNSWKNNKFSNPWCYENFIQARSLRRAQDIRKQMLGIMDRHKLDVVSCGKSTVRVQKAICSGFFRNAAKKDPQEGYRTLIDQQVVYIHPSSALFNRQPEWVVYHELVLTTKEYMREVTTIDPRWLVEFAPAFFKVSDPTKLSKQKKQQRLEPLYNRYEEPNAWRISRAFRRR | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1659 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction