Learn More
desmosome/hemidesmosome Human anti-Human, Clone: F12, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-81279-0.05MG
This item is not returnable.
View return policy
Description
desmosome/hemidesmosome Monoclonal specifically detects desmosome/hemidesmosome in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.
Specifications
| desmosome/hemidesmosome | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy | |
| Human | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunomicroscopy | |
| F12 | |
| Unconjugated | |
| PBS | |
| F12 was established by fusing peripheral blood lymphocytes derived from a human vulgaris patient with a hetermyeloma cell line. | |
| 0.05 mg | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgM κ |