missing translation for 'onlineSavingsMsg'
Learn More

desmosome/hemidesmosome Human anti-Human, Clone: F12, Novus Biologicals™

Human Monoclonal Antibody

463.00€ - 876.00€

Specifications

Antigen desmosome/hemidesmosome
Clone F12
Concentration 1 mg/mL
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy
Applications Flow Cytometry, Immunofluorescence, Immunomicroscopy
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18668051
View Documents
Novus Biologicals
NBP2-81279-25ug
25 μg
463.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18249839
View Documents
Novus Biologicals
NBP2-81279-0.05MG
0.05 mg
876.00€
0.05mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

desmosome/hemidesmosome Monoclonal specifically detects desmosome/hemidesmosome in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.
Specifications

Specifications

desmosome/hemidesmosome
1 mg/mL
Flow Cytometry, Immunofluorescence, Immunomicroscopy
Unconjugated
Human
Human
F12 was established by fusing peripheral blood lymphocytes derived from a human vulgaris patient with a hetermyeloma cell line.
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
F12
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy
Monoclonal
Purified
RUO
PBS
IgM κ
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.