Learn More
Desmoglein-4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 462.00€
Tekniske data
| Antigen | Desmoglein-4 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Beskrivelse
Desmoglein-4 Polyclonal antibody specifically detects Desmoglein-4 in Human, Mouse, Rat samples. It is validated for Western BlotTekniske data
| Desmoglein-4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 147409 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cadherin family member 13, CDGF13, CDHF13CDH family member 13, desmoglein 4, LAHdesmoglein-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 480-630 of human DSG4 (NP_817123.1). TGTICIEVPDINDYCPNIFPERRTICIDSPSVLISVNEHSYGSPFTFCVVDEPPGIADMWDVRSTNATSAILTAKQVLSPGFYEIPILVKDSYNRACELAQMVQLYACDCDDNHMCLDSGAAGIYTEDITGDTYGPVTEDQAGVSNVGLGP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel