Learn More
Desmoglein-4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92025-0.02ml
This item is not returnable.
View return policy
Description
Desmoglein-4 Polyclonal antibody specifically detects Desmoglein-4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Desmoglein-4 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000 | |
| Cadherin family member 13, CDGF13, CDHF13CDH family member 13, desmoglein 4, LAHdesmoglein-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 480-630 of human DSG4 (NP_817123.1). TGTICIEVPDINDYCPNIFPERRTICIDSPSVLISVNEHSYGSPFTFCVVDEPPGIADMWDVRSTNATSAILTAKQVLSPGFYEIPILVKDSYNRACELAQMVQLYACDCDDNHMCLDSGAAGIYTEDITGDTYGPVTEDQAGVSNVGLGP | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| 147409 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |