missing translation for 'onlineSavingsMsg'
Learn More

Delta5-Avenasterol (E/Z mixture), TRC

Product Code. 30007675
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007675 10 mg 10mg
30008143 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007675 Supplier Toronto Research Chemicals Supplier No. TRCA79482010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-36-1
Molecular Formula C29 H48 O
Molecular Weight (g/mol) 412.69
Synonym (3β)-Stigmasta-5,24(28)-dien-3-ol, Stigmasta-5,24(28)-dien-3β-ol, 24(28)-Ethylidenecholest-5-en-3β-ol, 24-Ethylcholesta-5,24(28)-dien-3β-ol, 24-Ethylidenecholest-5-en-3β-ol, 24-Ethylidenecholesterol, Stigmasta-5,24(28)-dien-3-ol, δ5-Avenasterin, δ5-Avenosterol
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-5-propan-2-ylhept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC=C(CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Analyte or Component Names delta5-Avenasterol (E,Z mixture)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h7,10,19-20,23-27,30H,8-9,11-18H2,1-6H3/t20-,23+,24+,25-,26+,27+,28+,29-/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 412.3705
Chemical Name or Material delta5-Avenasterol (E,Z mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.