CAS RN 18472-36-1
CAS RN 18472-36-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Delta5-Avenasterol (E/Z mixture), TRC
CAS: 18472-36-1 Molecular Formula: C29 H48 O Molecular Weight (g/mol): 412.69 Synonym: (3β)-Stigmasta-5,24(28)-dien-3-ol,Stigmasta-5,24(28)-dien-3β-ol,24(28)-Ethylidenecholest-5-en-3β-ol,24-Ethylcholesta-5,24(28)-dien-3β-ol,24-Ethylidenecholest-5-en-3β-ol,24-Ethylidenecholesterol,Stigmasta-5,24(28)-dien-3-ol,δ5-Avenasterin,δ5-Avenosterol IUPAC Name: (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-5-propan-2-ylhept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol SMILES: CC=C(CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C