missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Mouse anti-Human, Clone: CPTC-CSTA-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07781-100ug
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Cystatin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| Recombinant full-length human Cystatin A protein | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at 4°C. | |
| IgG2c κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CPTC-CSTA-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction