missing translation for 'onlineSavingsMsg'
Learn More

Cystatin A Mouse anti-Human, Clone: CPTC-CSTA-1, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen Cystatin A
Clone CPTC-CSTA-1
Concentration 0.2 mg/mL
Dilution Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655818
View Documents
Novus Biologicals
NBP3-07781-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cystatin A
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Tumor Suppressors
10 mM PBS with 0.05% BSA
1475
IgG2c κ
Protein A or G purified
CPTC-CSTA-1
Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A
Recombinant full-length human Cystatin A protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.