Learn More
Cyclopentanecarboxylic Acid, 98+%
Product Code. 10375740 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. 10375740
Brand: Thermo Scientific 111491000
Request Bulk or Custom Format
CAS: 3400-45-1 | C6H10O2 | 114.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3400-45-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00001371
InChI Key JBDSSBMEKXHSJF-UHFFFAOYSA-N
Synonym cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
PubChem CID 18840
IUPAC Name cyclopentanecarboxylic acid
SMILES C1CCC(C1)C(=O)O
Specifications
Melting Point 4.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 216.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CO2H
Refractive Index 1.4530 to 1.5450
Quantity 100 g
Beilstein 09, 6
Specific Gravity 1.05
Solubility Information Solubility in water: 3g/100mL. Other solubilities: soluble in alcohols, methylene chloride, acetone,, toluene
Formula Weight 114.14
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cyclopentanecarboxylic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.