missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin G Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92624-0.02ml
This item is not returnable.
View return policy
Description
Cyclin G Polyclonal antibody specifically detects Cyclin G in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Cyclin G | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| CCNG, CYCG1, cyclin G1, Cyclin-G, cyclin-G1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-295 of human Cyclin G (NP_004051.1). MIEVLTTTDSQKLLHQLNALLEQESRCQPKVCGLRLIESAHDNGLRMTARLRDFEVKDLLSLTQFFGFDTETFSLAVNLLDRFLSKMKVQPKHLGCVGLSCFYLAVKSIEEERNVPLATDLIRISQYRFTVSDLMRMEKIVLEKVCWKVKATTAFQFLQLYYSLLQENLPLERRNSINFERLEAQLKACHCRIIFSKAKPSVLALSIIALEIQAQKCVELTEGIECLQKHSKINGRDLTFWQELVSKCLTEYSSNKCSKPNVQKLKWIVSGRTARQLKHSYYRITHLPTIPEMVP | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 900 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction