Learn More
CstF-64 Mouse, anti-Human, Clone: 1C5G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68520-1-IG-20UL
 View more versions of this product
Product Code. 18030706
  • 145.00€ / 20µL
Description
The CstF-64 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CstF-64. This antibody recognizes human, mouse, rat antigen. The CstF-64 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CstF-64
1C5G2
CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2
20 μL
1478
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
150 kDa