missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CstF-64 Mouse, anti-Human, Clone: 1C5G2, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CstF-64 |
|---|---|
| Clone | 1C5G2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077035
|
Proteintech
68520-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030706
|
Proteintech
68520-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CstF-64 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CstF-64. This antibody recognizes human, mouse, rat antigen. The CstF-64 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CstF-64 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2 | |
| Primary |
| 1C5G2 | |
| Western Blot, ELISA | |
| Mouse | |
| 1478 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title