Learn More
CstF-64 Mouse, anti-Human, Clone: 1C5G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68520-1-IG-150UL
Description
The CstF-64 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CstF-64. This antibody recognizes human, mouse, rat antigen. The CstF-64 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CstF-64 | |
| 1C5G2 | |
| CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2 | |
| 150 μL | |
| 1478 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG1 |