missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
332.00€ - 481.00€
Specifications
| Antigen | CRISPR-Cas9 |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642889
|
Novus Biologicals
NBP3-05547-100ul |
100 μg |
481.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639847
|
Novus Biologicals
NBP3-05547-50ul |
50 μg |
332.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CRISPR-Cas9 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| Supplied as an aliquot of IgY preparation | |
| 901176 | |
| IgY | |
| IgY purified |
| Western Blot 1:1000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bacteria | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| C-terminal region of S. aureus CRISPR-Cas9 expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title