missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody, Novus Biologicals™

Chicken Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05547-50ul

 View more versions of this product

Product Code. 18639847

  • 332.00€ / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRISPR-Cas9
Polyclonal
Western Blot 1:1000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
C-terminal region of S. aureus CRISPR-Cas9 expressed in and purified from E. coli.
50 μg
Gene Editing, Gene Knockouts, Gene Therapy
901176
Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
IgY
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Supplied as an aliquot of IgY preparation
Chicken
IgY purified
RUO
Primary
Bacteria
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.