missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

170.00€ - 484.00€

Specifications

Antigen CRISPR-Cas9
Concentration 1.0mg/mL
Dilution Western Blot 2μg/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18266904
View Documents
Novus Biologicals
NBP2-66379SS
0.025 mg
170.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18268791
View Documents
Novus Biologicals
NBP2-66379
0.1 mg
484.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.
Specifications

Specifications

CRISPR-Cas9
Western Blot 2μg/ml
Polyclonal
Rabbit
Bacteria
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
IgG
Affinity Purified
Cas9 protein from Streptococcus pyogene serotype M1.
1.0mg/mL
Western Blot
Unconjugated
Gene Editing, Gene Knockouts, Gene Therapy
PBS with 0.02% Sodium Azide
Full length S. pyogenes Cas9 protein [UniProt Q99ZW2]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.