missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-66379
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.
Specifications
| CRISPR-Cas9 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| Cas9 protein from Streptococcus pyogene serotype M1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 2μg/ml | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| Full length S. pyogenes Cas9 protein [UniProt Q99ZW2] | |
| 0.1 mg | |
| Primary | |
| Bacteria | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction