missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
265.00€ - 629.00€
Specifications
| Classification | Monoclonal |
|---|---|
| Form | Purified |
| Isotype | IgG1 κ |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18770034
|
Novus Biologicals
NBP2-36440SS |
265.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18720914
|
Novus Biologicals
NBP2-36440 |
629.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, Immunoassay.Specifications
| Monoclonal | |
| Purified | |
| IgG1 κ | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title