Learn More
CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36440
 View more versions of this product
Product Code. 18720914
  • 629.00€ / 0.10mL
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Purified
IgG1 κ