Learn More
CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36440
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, Immunoassay.
Specifications
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| Purified | |
| IgG1 κ |