missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated SMN (Human,Mouse,Rat) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | SMN (Human,Mouse,Rat) |
|---|---|
| Clone | 3C10B5 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236982
|
Proteintech
CL488-60154-100UL |
100 μL |
450.00€ 349.00€ / 100µL Save 101.00€ 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234451
|
Proteintech
CL488-60154-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SMN (Human,Mouse,Rat) antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMN (Human,Mouse,Rat). This antibody recognizes human, mouse, rat antigen. The SMN (Human,Mouse,Rat) antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| SMN (Human,Mouse,Rat) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 6607 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3C10B5 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| BCD541; C-BCD541; FLJ76644; MGC20996; MGC5208; SMNC | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title