missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated SMN (Human,Mouse,Rat) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60154-20UL
This item is not returnable.
View return policy
Description
The SMN (Human,Mouse,Rat) antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMN (Human,Mouse,Rat). This antibody recognizes human, mouse, rat antigen. The SMN (Human,Mouse,Rat) antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| SMN (Human,Mouse,Rat) | |
| Monoclonal | |
| 1000 μg/mL | |
| BCD541; C-BCD541; FLJ76644; MGC20996; MGC5208; SMNC | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Conjugated Antibody | |
| IgG2b |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 3C10B5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6607 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction